CAS RN 910037-11-5
IUPAC Name:
2,3,4,5,6-pentafluorophenyl 3-(1-methyl-1H-pyrazol-3-yl)benzoate
Synonyms:
2,3,4,5,6-pentafluorophenyl 3-(1-methylpyrazol-3-yl)benzoate
Molecular Weight (g/mol):
368.26
Molecular Formula:
C17H9F5N2O2
InChi Key:
LGHVUZDAZTZCHC-UHFFFAOYSA-N
SMILES:
CN1C=CC(=N1)C1=CC(=CC=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
1
–
1
of
1
results
1
Pentafluorophenyl 3-(1-methyl-1H-pyrazol-3-yl)benzoate, 97%, Thermo Scientific™
CAS: 910037-11-5 Molecular Formula: C17H9F5N2O2 Molecular Weight (g/mol): 368.263 MDL Number: MFCD09065010 InChI Key: LGHVUZDAZTZCHC-UHFFFAOYSA-N Synonym: pentafluorophenyl 3-1-methyl-1h-pyrazol-3-yl benzoate,2,3,4,5,6-pentafluorophenyl 3-1-methylpyrazol-3-yl benzoate,pentafluorophenyl 3-1-methylpyrazol-3-yl benzoate,2,3,4,5,6-pentakis fluoranyl phenyl 3-1-methylpyrazol-3-yl benzoate,3-1-methyl-3-pyrazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester PubChem CID: 24229633 IUPAC Name: (2,3,4,5,6-pentafluorophenyl) 3-(1-methylpyrazol-3-yl)benzoate SMILES: CN1C=CC(=N1)C2=CC(=CC=C2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F