accessibility menu, dialog, popup

UserName

CH3NNFFFOOH

CAS RN 910037-16-0

IUPAC Name: 2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic acid
Synonyms: 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid
Molecular Weight (g/mol): 270.21
Molecular Formula: C12H9F3N2O2
InChi Key: ADEAPHCAMGDLFU-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CC=C1C(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results