accessibility menu, dialog, popup

UserName

H3COOClCl

CAS RN 91085-56-2

IUPAC Name: ethyl 3,5-dichlorobenzoate
Synonyms: ethyl 3,5-dichlorobenzoate
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: JRLNLVFPSMDPLU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results