accessibility menu, dialog, popup

UserName

CAS RN 912569-63-2

CH3NONN

CAS RN 912569-63-2

IUPAC Name: 4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazine-7-carbonitrile
Synonyms: 4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazine-7-carbonitrile
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: VLSVLSBIHZDQTF-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1N=CC(=C2)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results