missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbonitrile, 97%, Thermo Scientific™

Product Code. 11316855
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11316855 5 g Amber Glass Bottle 5g
10153174 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316855 Supplier Thermo Scientific Maybridge Supplier No. CC62016DE
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-63-2
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD09065019
InChI Key VLSVLSBIHZDQTF-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carbonitrile, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carbonitrile, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carbonitrile, 2h-pyrido 3,2-b-1,4-oxazine-7-carbonitrile,3,4-dihydro-4-methyl
PubChem CID 24229651
IUPAC Name 4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carbonitrile
SMILES CN1CCOC2=C1N=CC(=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.