accessibility menu, dialog, popup

UserName

NOFFFI

CAS RN 912761-82-1

IUPAC Name: 3-iodo-2-(2,2,2-trifluoroethoxy)pyridine
Synonyms: 3-iodo-2-(2,2,2-trifluoroethoxy)pyridine
Molecular Weight (g/mol): 303.02
Molecular Formula: C7H5F3INO
InChi Key: GUUWZDBHSBLCBA-UHFFFAOYSA-N
SMILES: FC(F)(F)COC1=C(I)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results