missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Iodo-2-(2,2,2-trifluoroethoxy)pyridine

Product Code. 11424550
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424550 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424550 Supplier Alfa Aesar™ Supplier No. H50108.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 912761-82-1
Molecular Formula C7H5F3INO
Molecular Weight (g/mol) 303.023
MDL Number MFCD07781168
InChI Key GUUWZDBHSBLCBA-UHFFFAOYSA-N
Synonym 3-iodo-2-2,2,2-trifluoroethoxy pyridine, 3-iodo-2-2,2,2-trifluoro-ethoxy-pyridine, acmc-20an2c
PubChem CID 42553135
IUPAC Name 3-iodo-2-(2,2,2-trifluoroethoxy)pyridine
SMILES C1=CC(=C(N=C1)OCC(F)(F)F)I

Specifications

Quantity 5 g
Sensitivity Light sensitive
Formula Weight 303.02
Chemical Name or Material 3-Iodo-2-(2,2,2-trifluoroethoxy)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.