accessibility menu, dialog, popup

UserName

CAS RN 913196-43-7

H3C(S)PNH2

CAS RN 913196-43-7

IUPAC Name: 1-[2-(diphenylphosphanyl)phenyl]ethan-1-amine
Synonyms: 1-[2-(diphenylphosphanyl)phenyl]ethanamine
Molecular Weight (g/mol): 305.36
Molecular Formula: C20H20NP
InChi Key: KPFJIPHGQGHIMM-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results