accessibility menu, dialog, popup

UserName

BNHOOH3CH3CCH3OOHHOOH

CAS RN 913835-72-0

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-5-(dihydroxyboranyl)benzoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-5-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol): 281.07
Molecular Formula: C12H16BNO6
InChi Key: DDZVPNHBJRCXEI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(=CC(=C1)C(O)=O)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results