accessibility menu, dialog, popup

UserName

CH3ONHHCl

CAS RN 916317-00-5

IUPAC Name: 4-(methoxymethyl)piperidine hydrochloride
Synonyms: 4-(methoxymethyl)piperidine hydrochloride
Molecular Weight (g/mol): 165.66
Molecular Formula: C7H16ClNO
InChi Key: FLRDPZJPRQWKLD-UHFFFAOYSA-N
SMILES: Cl.COCC1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results