missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxymethyl)piperidine hydrochloride

Product Code. 15447907
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447907 250 mg 250mg
11476690 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447907 Supplier Thermo Scientific Alfa Aesar Supplier No. H51020.MD
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 916317-00-5 | C7H16ClNO | 165.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916317-00-5
Molecular Formula C7H16ClNO
Molecular Weight (g/mol) 165.66
MDL Number MFCD08062602
InChI Key FLRDPZJPRQWKLD-UHFFFAOYSA-N
Synonym 4-methoxymethyl piperidine hydrochloride, 4-methoxymethyl piperidine hcl, 4-methoxymethyl-piperidine hydrochloride, 4-methoxymethylpiperidine hydrochloride, 4-methoxymethyl-piperidinehydrochloride, acmc-20a6g3, methoxymethylpiperidinehydrochloride, methoxy-4-piperidylmethane, chloride, 4-methoxymethyl piperidinehcl, 4-methoxymethyl piperidinehydrochloride
PubChem CID 17218599
IUPAC Name 4-(methoxymethyl)piperidine;hydrochloride
SMILES Cl.COCC1CCNCC1

Specifications

Quantity 250 mg
Formula Weight 165.66
Chemical Name or Material 4-(Methoxymethyl)piperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.