accessibility menu, dialog, popup

UserName

CH3OFFFOH

CAS RN 916420-98-9

IUPAC Name: [3-methoxy-5-(trifluoromethyl)phenyl]methanol
Synonyms: [3-methoxy-5-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: ROACFQURKLEGKL-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results