accessibility menu, dialog, popup

UserName

ON

CAS RN 92-71-7

IUPAC Name: 2,5-diphenyl-1,3-oxazole
Synonyms: 2,5-diphenyloxazole
Molecular Weight (g/mol): 221.26
Molecular Formula: C15H11NO
InChi Key: CNRNYORZJGVOSY-UHFFFAOYSA-N
SMILES: O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
Color 
  • (7)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results