CAS RN 921938-83-2
IUPAC Name:
2,3,4,5,6-pentafluorophenyl 4-methyl-2H,3H,4H-pyrido[3,2-b][1,4]oxazine-7-carboxylate
Synonyms:
2,3,4,5,6-pentafluorophenyl 4-methyl-2H,3H-pyrido[3,2-b][1,4]oxazine-7-carboxylate
Molecular Weight (g/mol):
360.24
Molecular Formula:
C15H9F5N2O3
InChi Key:
WEFDLXWSZQWZNH-UHFFFAOYSA-N
SMILES:
CN1CCOC2=C1N=CC(=C2)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
1
–
1
of
1
results
1
Pentafluorophenyl 4-methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylate, 97%, Thermo Scientific™
CAS: 921938-83-2 Molecular Formula: C15H9F5N2O3 Molecular Weight (g/mol): 360.24 MDL Number: MFCD09817508 InChI Key: WEFDLXWSZQWZNH-UHFFFAOYSA-N PubChem CID: 24229652 IUPAC Name: (2,3,4,5,6-pentafluorophenyl) 4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carboxylate SMILES: CN1CCOC2=C1N=CC(=C2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F