accessibility menu, dialog, popup

UserName

CAS RN 923281-52-1

BrOOFFF

CAS RN 923281-52-1

IUPAC Name: 5-bromo-2-(trifluoromethoxy)benzaldehyde
Synonyms: 5-bromo-2-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 269.02
Molecular Formula: C8H4BrF3O2
InChi Key: IBEWLWBZTHFCPI-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(C=O)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results