missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 15404377
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404377 10 g 10g
15494367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404377 Supplier Thermo Scientific Alfa Aesar Supplier No. H31751.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-(trifluoromethoxy)benzaldehyde is used in the synthesis of (-)-epicatechin, synthesis of biologically active compounds in vitro.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923281-52-1
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.02
MDL Number MFCD03701029
InChI Key IBEWLWBZTHFCPI-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethoxy benzaldehyde, benzaldehyde, 5-bromo-2-trifluoromethoxy, acmc-209rfd, 2-trifluoromethoxy-5-bromobenzaldehyde, 4-bromo-2-formyl-alpha,alpha,alpha-trifluoroanisole
PubChem CID 26599201
IUPAC Name 5-bromo-2-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=C(C=O)C=C(Br)C=C1

Specifications

Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 269.01
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-(trifluoromethoxy)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.