accessibility menu, dialog, popup

UserName

CAS RN 925-15-5

H3COOOOCH3

CAS RN 925-15-5

IUPAC Name: 1,4-dipropyl butanedioate
Synonyms: butanedioic acid, dipropyl ester
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: SZHZCPHKDJWHNG-UHFFFAOYSA-N
SMILES: CCCOC(=O)CCC(=O)OCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)