missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl succinate, 98%

Product Code. 11494397
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494397 10 g 10g
11404407 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494397 Supplier Thermo Scientific Alfa Aesar Supplier No. B20441.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 925-15-5 | C10H18O4 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-15-5
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00015213
InChI Key SZHZCPHKDJWHNG-UHFFFAOYSA-N
Synonym dipropyl succinate, di-n-propyl succinate, succinic acid, dipropyl ester, butanedioic acid, dipropyl ester, dipropylester kyseliny jantarove, dipropylester kyseliny jantarove czech, dipropyl butane-1,4-dioate, n-propyl succinate, 1,4-dipropyl butanedioate, succinic acid di-n-propyl ester
PubChem CID 13549
IUPAC Name dipropyl butanedioate
SMILES CCCOC(=O)CCC(=O)OCCC

Specifications

Density 1
Boiling Point 250°C to 252°C
Odor Pleasant
Quantity 10 g
Formula Weight 202.25
Percent Purity 98%
Chemical Name or Material Di-n-propyl succinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.