CAS RN 926268-51-1
CAS RN 926268-51-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-[(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)oxy]propanoic Acid, TRC
CAS: 926268-51-1 Molecular Formula: C13H16O4 Molecular Weight (g/mol): 236.26 IUPAC Name: 3-[(2,2-dimethyl-3H-1-benzofuran-7-yl)oxy]propanoic acid SMILES: CC1(C)Cc2cccc(OCCC(=O)O)c2O1