accessibility menu, dialog, popup

UserName

ClH2C

CAS RN 927-73-1

IUPAC Name: 4-chlorobut-1-ene
Synonyms: 4-chloro-1-butene
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: WKEVRZCQFQDCIR-UHFFFAOYSA-N
SMILES: ClCCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results