missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-butene, 98%, Thermo Scientific Chemicals

Product Code. 15562383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15562383 5 g 5g
15552383 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15562383 Supplier Thermo Scientific Acros Supplier No. 460160050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 927-73-1 | C4H7Cl | 90.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-73-1
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
InChI Key WKEVRZCQFQDCIR-UHFFFAOYSA-N
Synonym 4-chloro-1-butene, 1-butene, 4-chloro, 1-but-3-enyl chloride, ksc487m9b
PubChem CID 13565
IUPAC Name 4-chlorobut-1-ene
SMILES C=CCCCl

Specifications

Density 0.9210g/mL
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 0.921
Formula Weight 90.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-1-butene
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-1-butene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.