accessibility menu, dialog, popup

UserName

CH3NNClOCl

CAS RN 933-76-6

IUPAC Name: 4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one
Synonyms: 4,5-dichloro-2-methylpyridazin-3-one
Molecular Weight (g/mol): 179
Molecular Formula: C5H4Cl2N2O
InChi Key: ACKBTCUMGAHRIE-UHFFFAOYSA-N
SMILES: CN1N=CC(Cl)=C(Cl)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results