missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-methyl-3(2H)-pyridazinone, 97%, Thermo Scientific Chemicals

Product Code. 10743995 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10743995 5 g 5g
10506104 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10743995 Supplier Thermo Scientific Acros Supplier No. 399710050
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 933-76-6 | C5H4Cl2N2O | 179.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933-76-6
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179.00
MDL Number MFCD00051686
InChI Key ACKBTCUMGAHRIE-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-methylpyridazin-3 2h-one, 4,5-dichloro-2-methyl-3 2h-pyridazinone, 3 2h-pyridazinone, 4,5-dichloro-2-methyl, 4,5-dichloro-2-methyl-2h-pyridazin-3-one, 4,5-dichloro-2-methyl-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone,4,5-dichloro-2-methyl, 4,5-dichloro-2-methyl-2-hydropyridazin-3-one, pubchem16243, acmc-209wiz, 5-24-02-00023 beilstein handbook reference
PubChem CID 120462
IUPAC Name 4,5-dichloro-2-methylpyridazin-3-one
SMILES CN1N=CC(Cl)=C(Cl)C1=O

Specifications

Melting Point 87.0°C to 91.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 179.01
Percent Purity 97%
Chemical Name or Material 4, 5-Dichloro-2-methyl-3(2H)-pyridazinone
compliance-icons
Product Identifier
  • 4,5-Dichloro-2-methyl-3(2H)-pyridazinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.