accessibility menu, dialog, popup

UserName

H3COFF

CAS RN 93343-10-3

IUPAC Name: 1,3-difluoro-5-methoxybenzene
Synonyms: 1,3-difluoro-5-methoxybenzene
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: OTGQPYSISUUHAF-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC(F)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)