accessibility menu, dialog, popup

UserName

H3COHOHO

CAS RN 934-00-9

IUPAC Name: 3-methoxybenzene-1,2-diol
Synonyms: 1,2-benzenediol, 3-methoxy-
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: LPYUENQFPVNPHY-UHFFFAOYSA-N
SMILES: COC1=CC=CC(O)=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results