CAS RN 934246-14-7
CAS RN 934246-14-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Degarelix Acetate Hydrate, TRC
CAS: 934246-14-7 Molecular Formula: C82H103ClN18O16 . xC2H4O2 . xH2O Molecular Weight (g/mol): 1692.3 Synonym: N-acetyl-3-(2-naphthalenyl)-D-alanyl-4-chloro-D-phenylalanyl-3-(3-pyridinyl)-D-alanyl-L-seryl-4-[[[(4S)-hexahydro-2,6-dioxo-4-pyrimidinyl]carbonyl]amino]-L-phenylalanyl-4-[(aminocarbonyl)amino]-D-phenylalanyl-L-leucyl-N6-(1-methylethyl)-L-lysyl-L-prolyl-D-Alaninamide Acetate Hyrate SMILES: O.CC(C)C[C@H](NC(=O)[C@@H](Cc1ccc(NC(=O)N)cc1)NC(=O)[C@H](Cc2ccc(NC(=O)[C@H]3CC(=O)NC(=O)N3)cc2)NC(=O)[C@H](CO)NC(=O)[C@@H](Cc4cccnc4)NC(=O)[C@@H](Cc5ccc(Cl)cc5)NC(=O)[C@@H](Cc6ccc7ccccc7c6)NC(=O)C)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N8CCC[C@H]8C(=O)N[C@H](C)C(=O)N.CC(=O)O