missing translation for 'onlineSavingsMsg'
Learn More

Degarelix Acetate Hydrate, TRC

Product Code. 30374436
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374436 2.5 mg 2.5mg
30370191 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374436 Supplier Toronto Research Chemicals Supplier No. TRCD4954662.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934246-14-7
Molecular Formula C82H103ClN18O16 . xC2H4O2 . xH2O
Molecular Weight (g/mol) 1692.3
Synonym N-acetyl-3-(2-naphthalenyl)-D-alanyl-4-chloro-D-phenylalanyl-3-(3-pyridinyl)-D-alanyl-L-seryl-4-[[[(4S)-hexahydro-2,6-dioxo-4-pyrimidinyl]carbonyl]amino]-L-phenylalanyl-4-[(aminocarbonyl)amino]-D-phenylalanyl-L-leucyl-N6-(1-methylethyl)-L-lysyl-L-prolyl-D-Alaninamide Acetate Hyrate
SMILES O.CC(C)C[C@H](NC(=O)[C@@H](Cc1ccc(NC(=O)N)cc1)NC(=O)[C@H](Cc2ccc(NC(=O)[C@H]3CC(=O)NC(=O)N3)cc2)NC(=O)[C@H](CO)NC(=O)[C@@H](Cc4cccnc4)NC(=O)[C@@H](Cc5ccc(Cl)cc5)NC(=O)[C@@H](Cc6ccc7ccccc7c6)NC(=O)C)C(=O)N[C@@H](CCCCNC(C)C)C(=O)N8CCC[C@H]8C(=O)N[C@H](C)C(=O)N.CC(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C82H103ClN18O16.C2H4O2/c1-45(2)35-60(72(107)92-59(16-9-10-33-87-46(3)4)80(115)101-34-12-17-68(101)79(114)88-47(5)70(84)105)93-74(109)63(38-51-23-30-58(31-24-51)91-81(85)116)95-76(111)64(39-50-21-28-57(29-22-50)90-71(106)66-42-69(104)100-82(117)99-66)97-78(113)67(44-102)98-77(112)65(41-53-13-11-32-86-43-53)96-75(110)62(37-49-19-26-56(83)27-20-49)94-73(108)61(89-48(6)103)40-52-18-25-54-14-7-8-15-55(54)36-52;1-2(3)4/h7-8,11,13-15,18-32,36,43,45-47,59-68,87,102H,9-10,12,16-17,33-35,37-42,44H2,1-6H3,(H2,84,105)(H,88,114)(H,89,103)(H,90,106)(H,92,107)(H,93,109)(H,94,108)(H,95,111)(H,96,110)(H,97,113)(H,98,112)(H3,85,91,116)(H2,99,100,104,117);1H3,(H,3,4)/t47-,59+,60+,61-,62-,63-,64+,65-,66+,67+,68+;/m1./s1
Formula Weight 1708.7805
Chemical Name or Material Degarelix Acetate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.