accessibility menu, dialog, popup

UserName

SNNOOFFFFF

CAS RN 934570-42-0

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(thiomorpholin-4-yl)pyridine-4-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(thiomorpholin-4-yl)pyridine-4-carboxylate
Molecular Weight (g/mol): 390.33
Molecular Formula: C16H11F5N2O2S
InChi Key: CXWZKWSFJLNFMF-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC(=NC=C2)N2CCSCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results