accessibility menu, dialog, popup

UserName

NNNH2FFFBr

CAS RN 935534-47-7

IUPAC Name: 5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
Synonyms: 5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
Molecular Weight (g/mol): 242
Molecular Formula: C5H3BrF3N3
InChi Key: FAMGPURZLOTOKD-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C(=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results