missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine, 95%, Thermo Scientific™

Product Code. 15461689
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461689 5 g 5g
15438946 250 mg 250mg
15451689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461689 Supplier Thermo Scientific Alfa Aesar Supplier No. H64470.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935534-47-7
Molecular Formula C5H3BrF3N3
Molecular Weight (g/mol) 241.999
MDL Number MFCD09261254
InChI Key FAMGPURZLOTOKD-UHFFFAOYSA-N
Synonym 5-bromo-4-trifluoromethyl pyrimidin-2-amine, 5-bromo-4-trifluoromethyl-pyrimidin-2-ylamine, 5-bromo-4-trifluoromethyl-2-pyrimidinamine, ksc672m4h, 2-amino-5-bromo-4-trifluoromethylpyrimidine, 5-bromo-4-trifluoromethyl-2-pyrimidineamin, 5-bromo-4-trifluoromethyl-2-pyrimidylamine, 2-amino-5-bromo-4-trifluoromethyl pyrimidine, 2-pyrimidinamine, 5-bromo-4-trifluoromethyl, 5-bromo-4-trifluoromethyl pyrimidine-2-ylamine
PubChem CID 45382152
IUPAC Name 5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
SMILES C1=C(C(=NC(=N1)N)C(F)(F)F)Br

Specifications

Quantity 5 g
Formula Weight 242
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.