accessibility menu, dialog, popup

UserName

CAS RN 936-58-3

IUPAC Name: 1-phenylbut-3-en-1-ol
Synonyms: 1-phenylbut-3-en-1-ol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: RGKVZBXSJFAZRE-UHFFFAOYNA-N
SMILES: OC(CC=C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results