missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-buten-4-ol, 97%

Product Code. 15470308
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470308 25 g 25g
15460308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470308 Supplier Thermo Scientific Alfa Aesar Supplier No. H53466.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 936-58-3 | C10H12O | 148.205 g/mol

4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-58-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00039617
InChI Key RGKVZBXSJFAZRE-UHFFFAOYSA-N
Synonym 1-phenyl-3-buten-1-ol, 4-phenyl-1-buten-4-ol, 1-phenyl-but-3-en-1-ol, acmc-1bdtt, upenn_abs_025, 1-phenyl-3-buten-1-ol #, benzenemethanol,a-2-propen-1-yl, benzenemethanol, .alpha.-2-propenyl
PubChem CID 220119
IUPAC Name 1-phenylbut-3-en-1-ol
SMILES C=CCC(C1=CC=CC=C1)O

Specifications

Density 0.992
Boiling Point 234°C
Flash Point 103°C (217°F)
Refractive Index 1.53
Quantity 25 g
Formula Weight 148.2
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-buten-4-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.