accessibility menu, dialog, popup

UserName

CAS RN 93643-24-4

IUPAC Name: (8aR)-octahydropyrrolo[1,2-a]pyrazine
Synonyms: (8aR)-octahydropyrrolo[1,2-a]pyrazine
Molecular Weight (g/mol): 126.2
Molecular Formula: C7H14N2
InChi Key: FTTATHOUSOIFOQ-SSDOTTSWSA-N
SMILES: C1C[C@@H]2CNCCN2C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results