missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%

Product Code. 15498917
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498917 1 g 1g
15408927 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498917 Supplier Thermo Scientific Alfa Aesar Supplier No. H52295.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93643-24-4 | C7H14N2 | 126.20 g/mol

(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93643-24-4
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.20
MDL Number MFCD03787926
InChI Key FTTATHOUSOIFOQ-SSDOTTSWSA-N
Synonym s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
PubChem CID 781249
SMILES C1C[C@@H]2CNCCN2C1

Specifications

Quantity 1 g
UN Number UN3267
Solubility Information Slightly soluble in water.
IUPAC Name (8aR)-octahydropyrrolo[1,2-a]pyrazine
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material (S)-1,4-Diazabicyclo[4.3.0]nonane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.