accessibility menu, dialog, popup

UserName

CAS RN 937796-02-6

CH3HNSBr

CAS RN 937796-02-6

IUPAC Name: {[4-(3-bromothiophen-2-yl)phenyl]methyl}(methyl)amine
Synonyms: {[4-(3-bromothiophen-2-yl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 282.2
Molecular Formula: C12H12BrNS
InChi Key: JAQWWPQIUYWZIC-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)C1=C(Br)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results