accessibility menu, dialog, popup

UserName

CH3OHOCl

CAS RN 938-95-4

IUPAC Name: 2-(4-chlorophenyl)propanoic acid
Synonyms: 2-(4-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: YOZILQVNIWNPFP-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)