CAS RN 938184-94-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 3-(4-Cyano-4-(phenylamino)piperidin-1-yl)propanoate, TRC
CAS: 938184-94-2 Molecular Formula: C16H21N3O2 Molecular Weight (g/mol): 287.36 Synonym: Remifentanil Impurity 1 IUPAC Name: methyl 3-(4-anilino-4-cyanopiperidin-1-yl)propanoate SMILES: COC(=O)CCN1CCC(CC1)(Nc2ccccc2)C#N