missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-Cyano-4-(phenylamino)piperidin-1-yl)propanoate, TRC

Product Code. 30481968
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481968 100 mg 100mg
30477659 50 mg 50mg
30465383 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481968 Supplier Toronto Research Chemicals Supplier No. TRCM218856100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938184-94-2
Molecular Formula C16H21N3O2
Molecular Weight (g/mol) 287.36
Synonym Remifentanil Impurity 1
IUPAC Name methyl 3-(4-anilino-4-cyanopiperidin-1-yl)propanoate
SMILES COC(=O)CCN1CCC(CC1)(Nc2ccccc2)C#N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H21N3O2/c1-21-15(20)7-10-19-11-8-16(13-17,9-12-19)18-14-5-3-2-4-6-14/h2-6,18H,7-12H2,1H3
Formula Weight 287.1634
Chemical Name or Material Methyl 3-(4-Cyano-4-(phenylamino)piperidin-1-yl)propanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.