accessibility menu, dialog, popup

UserName

CAS RN 939-83-3

CH3NNOO

CAS RN 939-83-3

IUPAC Name: 2-methyl-5-nitrobenzonitrile
Synonyms: 2-methyl-5-nitrobenzonitrile
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: XOSDYLFXPMFRGF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C#N)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results