accessibility menu, dialog, popup

UserName

CAS RN 93917-68-1

H3COOOH3C

CAS RN 93917-68-1

IUPAC Name: ethyl 2-(2-methylphenoxy)acetate
Synonyms: ethyl 2-(2-methylphenoxy)acetate
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: NHFQSFIBSJPLIW-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=CC=CC=C1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results