accessibility menu, dialog, popup

UserName

CAS RN 939807-75-7

BOOH3CCH3H3CH3CFNH2F

CAS RN 939807-75-7

IUPAC Name: 2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 255.07
Molecular Formula: C12H16BF2NO2
InChi Key: SOHBHFWRCLJGDV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results