missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester, 96%

Product Code. 15419755
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419755 250 mg 250mg
15409755 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15419755 Supplier Thermo Scientific Alfa Aesar Supplier No. H52356.MD
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 939807-75-7 | C12H16BF2NO2 | 255.072 g/mol

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939807-75-7
Molecular Formula C12H16BF2NO2
Molecular Weight (g/mol) 255.072
MDL Number MFCD09743045
InChI Key SOHBHFWRCLJGDV-UHFFFAOYSA-N
Synonym 4-amino-2,5-difluorobenzeneboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine, 4-amino-2,5-difluorophenylboronic acid pinacol ester, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 57955144
IUPAC Name 2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)N)F

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 255.07
Percent Purity 96%
Chemical Name or Material 4-Amino-2,5-difluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.