accessibility menu, dialog, popup

UserName

CAS RN 94-18-8

IUPAC Name: benzyl 4-hydroxybenzoate
Synonyms: benzylparaben benzyl parasept benzyl hydroxybenzoate benzyl tegosept benzyl-parasept nisapulvol solbrol Z benzyl parahydroxybenzoate nipabenzyl parosept
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: MOZDKDIOPSPTBH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results