accessibility menu, dialog, popup

UserName

CAS RN 94-92-8

IUPAC Name: N1,N2-bis(2-methylphenyl)ethane-1,2-diamine
Synonyms: N1,N2-bis(2-methylphenyl)ethane-1,2-diamine
Molecular Weight (g/mol): 240.35
Molecular Formula: C16H20N2
InChi Key: ZQMPWXFHAUDENN-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NCCNC1=CC=CC=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results