accessibility menu, dialog, popup

UserName

CAS RN 94-96-2

H3CHOCH3OH

CAS RN 94-96-2

IUPAC Name: 2-ethylhexane-1,3-diol
Synonyms: 2-ethyl-1,3-hexanediol compound 6-12 insect repellent caswell no. 445 ethyl hexanediol (substance) epa pesticide chemical code 041001 ethohexadiol latka 612 ethohexadiol, (threo)- carbide 6-12 etohexadiol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: RWLALWYNXFYRGW-UHFFFAOYNA-N
SMILES: CCCC(O)C(CC)CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)