missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexane-1,3-diol, 97%

Product Code. 11464263
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464263 10,000 mL 10000mL
11444263 500 mL 500mL
11454263 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464263 Supplier Thermo Scientific Alfa Aesar Supplier No. A10898.0D
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-96-2 | C8H18O2 | 146.23 g/mol

2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

Solubility
Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-96-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004578
InChI Key RWLALWYNXFYRGW-UHFFFAOYNA-N
Synonym 2-ethyl-1,3-hexanediol, ethohexadiol, 1,3-hexanediol, 2-ethyl, octylene glycol, ethyl hexanediol, carbide 6-12, repellent 612, rutgers 612, 6-12-insect repellent, diol-kyowa 8
PubChem CID 7211
ChEBI CHEBI:34273
IUPAC Name 2-ethylhexane-1,3-diol
SMILES CCCC(O)C(CC)CO

Specifications

Melting Point -40°C
Boiling Point 241°C to 249°C
Odor Odorless
Linear Formula CH3CH2CH2CHOHCH(CH2CH3)CH2OH
Quantity 10,000 mL
Solubility Information Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 2-Ethylhexane-1,3-diol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.