accessibility menu, dialog, popup

UserName

NClNF

CAS RN 942206-11-3

IUPAC Name: 5-chloro-2'-fluoro-2,4'-bipyridine
Synonyms: 5-chloro-2'-fluoro-2,4'-bipyridine
Molecular Weight (g/mol): 208.62
Molecular Formula: C10H6ClFN2
InChi Key: UCTHNJGQZMBIOJ-UHFFFAOYSA-N
SMILES: FC1=NC=CC(=C1)C1=NC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results