accessibility menu, dialog, popup

UserName

CAS RN 94413-64-6

CH3OONN

CAS RN 94413-64-6

IUPAC Name: methyl 2-cyanopyridine-4-carboxylate
Synonyms: methyl 2-cyanopyridine-4-carboxylate
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: ORVHMLCJEKDDAX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=NC=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results