missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-cyanoisonicotinate, 97%, Thermo Scientific™

Product Code. 15429588
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15429588

Brand: Thermo Scientific Alfa Aesar H63090.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 2-cyanoisonicotinate can be used in the synthesis of various pharmaceutical and biologically active compounds, such as tetra substituted imidazolines as potent and selective neuropeptide Y (NPY) Y5 receptor antagonists.

Solubility
Slightly Soluble in water (6.2 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94413-64-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD07367894
InChI Key ORVHMLCJEKDDAX-UHFFFAOYSA-N
Synonym methyl 2-cyanoisonicotinate, 2-cyano-4-pyridine carboxylic acid methyl ester, 4-pyridinecarboxylicacid, 2-cyano-, methyl ester, 4-pyridinecarboxylic acid, 2-cyano-, methyl ester, methyl 2-cyanois, methyl 2-cyanoisonicotite, 2-cyano-4-carbomethoxypyridine, 2-cyanoisonicotinic acid methyl ester, 2-cyano-isonicotinic acid methyl ester, 2-cyano-4-pyridine-carboxylic acid methyl ester
PubChem CID 12132796
IUPAC Name methyl 2-cyanopyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)C#N

Specifications

Color Brown
Quantity 250 mg
UN Number UN3439
Solubility Information Slightly Soluble in water (6.2g/L) (25°C).
Formula Weight 162.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-cyanoisonicotinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.