accessibility menu, dialog, popup

UserName

CAS RN 944450-92-4

NClOHCl

CAS RN 944450-92-4

IUPAC Name: 4-(chloromethyl)-1-[(furan-2-yl)methyl]piperidine hydrochloride
Synonyms: 4-(chloromethyl)-1-(furan-2-ylmethyl)piperidine hydrochloride
Molecular Weight (g/mol): 250.16
Molecular Formula: C11H17Cl2NO
InChi Key: SRRFZUHMIKVVPV-UHFFFAOYSA-N
SMILES: Cl.ClCC1CCN(CC2=CC=CO2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results